missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)aniline, 97%

Product Code. 10623805 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10623805 5 g Glass bottle 5g
10180744 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10623805 Supplier Thermo Scientific Acros Supplier No. 127110050
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket
This item is not returnable. View return policy

CAS: 2987-53-3 | C7H9NS | 139.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2987-53-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.22
MDL Number MFCD00007708
InChI Key WBRPQQSADOCKCH-UHFFFAOYSA-N
Synonym 2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline
PubChem CID 76337
IUPAC Name 2-methylsulfanylaniline
SMILES CSC1=CC=CC=C1N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Density 1.1110g/mL
Boiling Point 234.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3SC6H4NH2
Refractive Index 1.6230 to 1.6250 (20°C, 589nm)
Quantity 5 g
Beilstein 13, 399
Specific Gravity 1.111
Formula Weight 139.22
Percent Purity 97%
Physical Form Chunks and/or Powder
Chemical Name or Material 2-(Methylthio)aniline
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Methylthio)aniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.