missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)aniline, 98%

Product Code. 11495093
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495093 100 g 100g
11475093 5 g 5g
11485093 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495093 Supplier Thermo Scientific Alfa Aesar Supplier No. A12762.22
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 2987-53-3 | C7H9NS | 139.216 g/mol

2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)aniline used as important raw material and intermediate used in organic Synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water (less).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2987-53-3
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007708
InChI Key WBRPQQSADOCKCH-UHFFFAOYSA-N
Synonym 2-methylthio aniline, 2-aminothioanisole, 2-methylsulfanyl aniline, 2-methylmercaptoaniline, 2-methylmercapto aniline, o-thioanisidine, o-aminothioanisole, 2-amino thioanisole, benzenamine, 2-methylthio, o-methylthio aniline
PubChem CID 76337
IUPAC Name 2-methylsulfanylaniline
SMILES CSC1=CC=CC=C1N

Specifications

Density 1.111
Boiling Point 233°C to 234°C
Flash Point >110°C (230°F)
Refractive Index 1.624
Quantity 100 g
UN Number UN2810
Beilstein 386211
Solubility Information Soluble in water (less).
Formula Weight 139.23
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.