missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzonitrile, 98%

Product Code. 11367026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11367026 5 g 5g
11357026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367026 Supplier Thermo Scientific Alfa Aesar Supplier No. L06870.06
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6609-54-7 | C8H7NS | 149.211 g/mol

2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylthio)benzonitrile is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6609-54-7
Molecular Formula C8H7NS
Molecular Weight (g/mol) 149.211
MDL Number MFCD00015557
InChI Key PXZSANDJGNKIIA-UHFFFAOYSA-N
Synonym 2-methylthio benzonitrile, 2-methylsulfanyl benzonitrile, 2-cyanophenyl methyl sulfide, 2-methylmercapto benzonitrile, methylthio benzonitrile, 2-methylthiobenzenecarbonitrile, benzonitrile,2-methylthio, 2-methylsulfanyl benzenecarbonitrile
PubChem CID 138781
IUPAC Name 2-methylsulfanylbenzonitrile
SMILES CSC1=CC=CC=C1C#N

Specifications

Melting Point 33°C to 38°C
Boiling Point 116°C to 117°C (5 mmHg)
Quantity 5 g
UN Number UN3439
Beilstein 2207135
Solubility Information Insoluble in water.
Formula Weight 149.22
Percent Purity 98%
Chemical Name or Material 2-(Methylthio)benzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.