missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)benzylamine, 95%, Thermo Scientific™

Product Code. 10683793
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10683793 1 g 1g
10374482 250 mg 250mg
10407564 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10683793 Supplier Thermo Scientific Maybridge Supplier No. KM09736DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 56004-83-2
Molecular Formula C8H11NS
Molecular Weight (g/mol) 153.243
MDL Number MFCD00831039
InChI Key YVEJLBIEESKJLG-UHFFFAOYSA-N
Synonym 2-methylthio benzylamine, 2-methylsulfanyl phenyl methanamine, 2-methylthio phenyl methanamine, 1-2-methylsulfanyl phenyl methanamine, 2-methylsulfanylphenyl methanamine, 2-methylthiobenzylamine, 2-methylthio benzyl amine, 2-methylthio-benzyl amine, 2-methylthiophenyl methylamine
PubChem CID 2736981
IUPAC Name (2-methylsulfanylphenyl)methanamine
SMILES CSC1=CC=CC=C1CN

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber glass bottle
Linear Formula CH3SC6H4CH2NH2
Quantity 1 g
Formula Weight 153.25
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.