missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylthio)ethanol, 99%

Product Code. 10511282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10511282 5 mL Glass bottle 5mL
10328110 25 mL Glass bottle 25mL
10761535 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10511282 Supplier Thermo Scientific Acros Supplier No. 210100050
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5271-38-5
Molecular Formula C3H8OS
Molecular Weight (g/mol) 92.16
MDL Number MFCD00002908
InChI Key WBBPRCNXBQTYLF-UHFFFAOYSA-N
Synonym 2-methylthio ethanol, methylthioethanol, ethanol, 2-methylthio, 2-hydroxyethyl methyl sulfide, 2-methylmercaptoethanol, 2-methylsulfanyl ethanol, 2-methylsulfanyl-ethanol, beta-hydroxyethyl methyl sulfide, 2-methylthioethanol
PubChem CID 78925
ChEBI CHEBI:63861
IUPAC Name 2-methylsulfanylethanol
SMILES CSCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.0600g/mL
Boiling Point 169°C to 171°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3SCH2CH2OH
Refractive Index 1.4934 to 1.4954
Quantity 5 mL
Beilstein 01, II, 524
Specific Gravity 1.06
Solubility Information Solubility in water: insoluble
Formula Weight 92.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylthio)ethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Methylthio)ethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.