missing translation for 'onlineSavingsMsg'
Learn More

2-Methylthiophene, 98%

Product Code. 10001300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10001300 25 g Glass bottle 25g
10066193 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10001300 Supplier Thermo Scientific Acros Supplier No. 127850250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 554-14-3 | C5H6S | 98.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 554-14-3
Molecular Formula C5H6S
Molecular Weight (g/mol) 98.16
MDL Number MFCD00005451
InChI Key XQQBUAPQHNYYRS-UHFFFAOYSA-N
Synonym thiophene, 2-methyl, 2-methyl thiophene, thiophene, methyl, alpha-methylthiophene, ccris 2936, 2-methyl-thiophene, unii-7115jap77a, 2-methylthiacyclopentadiene, a-thiotolene, pubchem5204
PubChem CID 11126
IUPAC Name 2-methylthiophene
SMILES CC1=CC=CS1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -63°C
Color Colorless to Yellow
Density 1.0200g/mL
Boiling Point 113°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5190 to 1.521
Quantity 25 g
Beilstein 17, 37
Specific Gravity 1.02
Solubility Information Solubility in water: immiscible
Formula Weight 98.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methylthiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methylthiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.