missing translation for 'onlineSavingsMsg'
Learn More

2-Morpholinoisonicotinaldehyde, 97%, Thermo Scientific™

Product Code. 10095772
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095772 250 mg Amber Glass Bottle 250mg
10249342 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10095772 Supplier Thermo Scientific Maybridge Supplier No. CC43104CB
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 864068-87-1
Molecular Formula C10H12N2O2
Molecular Weight (g/mol) 192.22
MDL Number MFCD08060524
InChI Key VMFCHBZYIHUMQE-UHFFFAOYSA-N
PubChem CID 7537563
SMILES O=CC1=CC(=NC=C1)N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2-morpholinoisonicotinaldehyde, 2-morpholin-4-yl pyridine-4-carbaldehyde, 2-morpholin-4-yl pyridine-4-carboxaldehyde, 4-pyridinecarboxaldehyde,2-4-morpholinyl, 2-morpholin-4-yl isonicotinaldehyd, 2-morpholin-4-yl-pyridine-4-carbaldehyde, 4-pyridinecarboxaldehyde, 2-4-morpholinyl
IUPAC Name 2-(morpholin-4-yl)pyridine-4-carbaldehyde
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material 2-Morpholinoisonicotinaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.