missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinomethyl)benzaldehyde 97%, Thermo Scientific™

Product Code. 10554004
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10554004 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10554004 Supplier Thermo Scientific Maybridge Supplier No. CC48604CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 736991-21-2
Assay Percent Range 97%
Molecular Formula C12H15NO2
MDL Number MFCD07782412
Quantity 250 mg
Synonym 2-morpholinomethyl benzaldehyde, 2-morpholin-4-ylmethyl benzaldehyde, morpholinomethylbenzaldehyde, 4-2-carboxaldehydebenzyl morpholine, 2-morpholin-4-yl methyl benzaldehyde
InChI Key YPZNTOVVELIJGI-UHFFFAOYSA-N
SMILES O=CC1=CC=CC=C1CN1CCOCC1
IUPAC Name 2-[(morpholin-4-yl)methyl]benzaldehyde
Molecular Weight (g/mol) 205.26
PubChem CID 18525826
Formula Weight 205.26
Chemical Name or Material 2-(morpholinomethyl)benzaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.