missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinosulfonyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 10682285
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10682285 250 mg Amber Glass Bottle 250mg
10275314 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10682285 Supplier Thermo Scientific Maybridge Supplier No. CC69604CB
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 862500-24-1
Molecular Formula C11H13NO4S
Molecular Weight (g/mol) 255.288
MDL Number MFCD09817561
InChI Key GSDGTTZDQFKUBA-UHFFFAOYSA-N
Synonym 2-morpholinosulfonyl benzaldehyde, 2-morpholine-4-sulfonyl benzaldehyde, 2-morpholin-4-ylsulfonyl benzaldehyde, 2-morpholin-4-ylsulphonyl benzaldehyde, benzaldehyde,2-4-morpholinylsulfonyl
PubChem CID 24229770
IUPAC Name 2-morpholin-4-ylsulfonylbenzaldehyde
SMILES C1COCCN1S(=O)(=O)C2=CC=CC=C2C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 255.29
Percent Purity 97%
Chemical Name or Material 2-(Morpholinosulfonyl)benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.