missing translation for 'onlineSavingsMsg'
Learn More

2-(morpholinosulfonyl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10777865
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10777865 1 g Amber Glass Bottle 1g
10152984 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10777865 Supplier Thermo Scientific Maybridge Supplier No. CC69601DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 87223-34-5
Molecular Formula C11H13NO5S
Molecular Weight (g/mol) 271.287
MDL Number MFCD09813541
InChI Key YTQKWJNSSYODEV-UHFFFAOYSA-N
Synonym 2-morpholinosulfonyl benzoic acid, 2-morpholine-4-sulfonyl benzoic acid, 2-morpholin-4-ylsulfonyl benzoic acid, benzoic acid,2-4-morpholinylsulfonyl, 2-morpholin-4-ylsulphonyl benzoic acid
PubChem CID 13140207
IUPAC Name 2-morpholin-4-ylsulfonylbenzoic acid
SMILES C1COCCN1S(=O)(=O)C2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 271.29
Percent Purity 97%
Chemical Name or Material 2-(Morpholinosulfonyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.