missing translation for 'onlineSavingsMsg'
Learn More

2-(Morpholinosulfonyl)benzylamine hydrochloride, 97%, Thermo Scientific™

Product Code. 10657864
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657864 250 mg Amber Glass Bottle 250mg
10235994 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10657864 Supplier Thermo Scientific Maybridge Supplier No. CC69663CB
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 918812-18-7
Molecular Formula C11H17ClN2O3S
Molecular Weight (g/mol) 292.778
MDL Number MFCD09284586
InChI Key RMMDNVBLYNZQFP-UHFFFAOYSA-N
Synonym 2-morpholinosulfonyl benzylamine hydrochloride, 1-2-morpholin-4-ylsulfonyl phenyl methanamine hydrochloride, 2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 1-2-morpholine-4-sulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride, 2-morpholinosulfonyl phenyl methanamine hydrochloride salt, 1-2-morpholine-4-sulfonyl phenyl methanamine-hydrogen chloride 1/1
PubChem CID 16320333
IUPAC Name (2-morpholin-4-ylsulfonylphenyl)methanamine;hydrochloride
SMILES C1COCCN1S(=O)(=O)C2=CC=CC=C2CN.Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 292.79
Percent Purity 97%
Chemical Name or Material 2-(Morpholinosulfonyl)benzylamine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.