missing translation for 'onlineSavingsMsg'
Learn More

[2-(Morpholinosulfonyl)phenyl]methanol, ≥97%, Thermo Scientific™

Product Code. 10622675
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10622675 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10622675 Supplier Thermo Scientific Maybridge Supplier No. CC69609DA
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 937796-15-1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Molecular Formula C11H15NO4S
MDL Number MFCD09817562
Quantity 1 g
Synonym 2-morpholinosulfonyl phenyl methanol, 2-morpholine-4-sulfonyl phenyl methanol, 2-4-morpholinylsulfonyl phenyl methanol, 2-morpholinosulfonyl phenyl methanol,97+%, 4-2-hydroxymethyl phenyl sulfonyl morpholine
InChI Key BOQHDNNWBAICEW-UHFFFAOYSA-N
SMILES OCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
IUPAC Name (2-morpholin-4-ylsulfonylphenyl)methanol
Molecular Weight (g/mol) 257.30
PubChem CID 24229772
Formula Weight 257.31
Percent Purity 97%
Chemical Name or Material [2-(Morpholinosulfonyl)phenyl]methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.