missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenesulfonic acid hydrate, 90%, Technical

Product Code. 10486169 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10486169 5 g Glass bottle 5g
10187949 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10486169 Supplier Thermo Scientific Acros Supplier No. 428690050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 76530-12-6 | C10H8O3S | 208.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76530-12-6 , 120-18-3
Molecular Formula C10H8O3S
Molecular Weight (g/mol) 208.23
MDL Number MFCD00149271
InChI Key KVBGVZZKJNLNJU-UHFFFAOYSA-N
Synonym naphthalene-2-sulfonic acid hydrate, 2-naphthalenesulfonic acid hydrate, 2-naphthalenesulfonic acid monohydrate, 2-naphthalenesulfonicacid, hydrate 1:1, kyselina2-naftalensulfonova, acmc-20aj4s, ksc352o8d, c10h8so3.h2o, 2-naphthalene-sulfonic acid monohydrate, naphthalene-2-sulphonic acid monohydrate
PubChem CID 20499058
IUPAC Name naphthalene-2-sulfonic acid;hydrate
SMILES OS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color Gray
Density 1.4400g/mL
Infrared Spectrum Authentic
Assay Percent Range 88% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 1.44
Solubility Information Solubility in water: soluble
Formula Weight 208.24
Percent Purity 90%
Grade Technical
Physical Form Powder
Chemical Name or Material 2-Naphthalenesulfonic acid hydrate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Naphthalenesulfonic acid hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.