missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenesulfonyl chloride, 97%

Product Code. 10456810 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10456810

Brand: Thermo Scientific Acros 158440250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 93-11-8 | C10H7ClO2S | 226.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 93-11-8
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.67
MDL Number MFCD00004087
InChI Key OPECTNGATDYLSS-UHFFFAOYSA-N
Synonym 2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
PubChem CID 7125
IUPAC Name naphthalene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Spécification

CAS Min % 96.0
CAS Max % 100.0
Melting Point 74°C to 78°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C10H7SO2Cl
Quantity 25 g
Beilstein 11, 173
Solubility Information Solubility in water: insoluble
Formula Weight 226.68
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Naphthalenesulfonyl chloride
Afficher plus Afficher moins
compliance-icons compliance-icons
Identifiant produit
  • 2-Naphthalenesulfonyl chloride
Mention d'avertissement
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
Mention de danger
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H261-In contact with water releases flammable gases.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Correction du contenu d'un produit

Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.

Nom du produit

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.

Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis