missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenethiol, 98+%

Product Code. 10643762 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10643762 25 g Glass bottle 25g
10569531 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10643762 Supplier Thermo Scientific Acros Supplier No. 334520250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 91-60-1 | C10H8S | 160.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 91-60-1
Molecular Formula C10H8S
Molecular Weight (g/mol) 160.2
MDL Number MFCD00004086
InChI Key RFCQDOVPMUSZMN-UHFFFAOYSA-N
Synonym 2-naphthalenethiol, 2-mercaptonaphthalene, 2-thionaphthol, thionaphthol, 2-naphthyl mercaptan, vulcamel tbn, 2-naphthyl thiol, renacit 1, rpa no. 2, rpa 2
PubChem CID 7058
IUPAC Name naphthalene-2-thiol
SMILES C1=CC=C2C=C(C=CC2=C1)S

Specifikationer

CAS Min % 98.5
CAS Max % 100.0
Melting Point 79°C to 83°C
Color Cream to White
Boiling Point 286°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 6464
Solubility Information Solubility in water: .. Other solubilities: soluble in organic solvents
Formula Weight 160.2
Percent Purity 98+%
Physical Form Powder
Chemical Name or Material 2-Naphthalenethiol, 99%
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • 2-Naphthalenethiol
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
Faroangivelser
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Skyddsangivelse
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.