missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoic acid, 98+%

Product Code. p-7044189
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11371827

Brand: Thermo Scientific Alfa Aesar L05102.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 93-09-4 | C11H8O2 | 172.183 g/mol

2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

Solubility
Soluble in alcohol, diethyl ether, methanol, and ethyl acetate (20 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-09-4
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004101
InChI Key UOBYKYZJUGYBDK-UHFFFAOYSA-N
Synonym 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
PubChem CID 7123
ChEBI CHEBI:36106
IUPAC Name naphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)O

Specifications

Melting Point 182°C to 186°C
Density 1.08
Flash Point 205°C (401°F)
Quantity 5 g
Beilstein 972039
Merck Index 14,6382
Solubility Information Soluble in alcohol,diethyl ether,methanol,and ethyl acetate (20mg/ml). Insoluble in water.
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material 2-Naphthoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.