missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoic acid, 99%

Product Code. 10347500 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10347500 25 g Glass bottle 25g
15700829 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10347500 Supplier Thermo Scientific Acros Supplier No. 177320250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 93-09-4 | C11H8O2 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-09-4
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.18
MDL Number MFCD00004101
InChI Key UOBYKYZJUGYBDK-UHFFFAOYSA-N
Synonym 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
PubChem CID 7123
ChEBI CHEBI:36106
IUPAC Name naphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 183°C to 187°C
Color White to Beige
Density 205g/mL
Boiling Point >300°C
Flash Point 205°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C10H7CO2H
Quantity 25 g
Beilstein 09, 656
Merck Index 15, 6467
Solubility Information Solubility in water: <0.5g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 172.18
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 2-Naphthoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Naphthoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.