missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoxyacetic acid, 97%

Product Code. 11488386
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488386 100 g 100g
11498386 500 g 500g
11408396 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11488386 Supplier Thermo Scientific Alfa Aesar Supplier No. A15412.22
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 120-23-0 | C12H10O3 | 202.209 g/mol

2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoxyacetic acid is a phytohormone and plays a vital role in plant growth regulator on tomatoes and strawberries.

Solubility
Soluble in alcohol, ether, acetic acid and dimethyl sulfoxide. Slightly soluble in water.

Notes
Store in a cool place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-23-0
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004066
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
PubChem CID 8422
ChEBI CHEBI:50397
IUPAC Name 2-naphthalen-2-yloxyacetic acid
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O

Specifications

Melting Point 155°C to 157°C
Quantity 100 g
Beilstein 1074148
Merck Index 14,6397
Solubility Information Soluble in alcohol,ether,acetic acid and dimethyl sulfoxide. Slightly soluble in water.
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 2-Naphthoxyacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.