missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl phenyl ketone, 98%, Thermo Scientific™

Product Code. 10567742 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10567742

Brand: Thermo Scientific Acros 415420050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 644-13-3
Molecular Formula C17H12O
Molecular Weight (g/mol) 232.28
MDL Number MFCD00004106
InChI Key SJNXJRVDSTZUFB-UHFFFAOYSA-N
Synonym 2-benzoylnaphthalene, 2-naphthyl phenyl ketone, 2-benzonaphthone, naphthalen-2-yl phenyl methanone, methanone, 2-naphthalenylphenyl, 2'-benzonaphthone, ketone, 2-naphthyl phenyl, 2-naphthylphenylketone, beta-benzoylnaphthalene, 2-naphthyl phenyl methanone
PubChem CID 69516
IUPAC Name naphthalen-2-yl(phenyl)methanone
SMILES O=C(C1=CC=CC=C1)C1=CC=C2C=CC=CC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76°C to 82°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 232.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Naphthyl phenyl ketone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.