missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethoxy)aniline, 95%, Thermo Scientific™

Product Code. p-4510610
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10495332

Brand: Thermo Scientific Maybridge TL00377DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 2267-23-4
Molecular Formula C7H5F3N2O3
Molecular Weight (g/mol) 222.12
MDL Number MFCD00042326
InChI Key YCGFVAPIBALHRT-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethoxy aniline, 2-amino-5-trifluoromethoxy nitrobenzene, benzenamine, 2-nitro-4-trifluoromethoxy, 1-amino-2-nitro-4-trifluoromethoxy benzene, 2-nitro-4-trifluoromethoxy phenylamine, 4-amino-3-nitro-alpha,alpha,alpha-trifluoroanisole, pubchem8514, acmc-1cnwp, ksc496g9b, 2-nitro4-trifluoromethoxyaniline
PubChem CID 2775772
IUPAC Name 2-nitro-4-(trifluoromethoxy)aniline
SMILES NC1=CC=C(OC(F)(F)F)C=C1[N+]([O-])=O

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 222.12
Percent Purity 95%
Chemical Name or Material 2-Nitro-4-(trifluoromethoxy)aniline
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.