missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15770859 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15770859 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15770859 Supplier Thermo Scientific Acros Supplier No. 348110010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 320-94-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 137.0°C to 140.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C8H4F3NO4
Quantity 1 g
Synonym 2-nitro-4-trifluoromethyl benzoic acid, 2-nitro-4-trifluoromethylbenzoic acid, ntfa, 4-trifluoromethyl-2-nitrobenzoic acid, benzoic acid, 2-nitro-4-trifluoromethyl, unii-k9m35sv4p3, 2-nitro-4-trifluoromethyl-benzoic acid, pubchem4539, acmc-1ahai, 2ntf-boa
InChI Key MYSAXQPTXWKDPQ-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(=O)O
Molecular Weight (g/mol) 235.11
PubChem CID 642096
Formula Weight 235.11
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzoic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.