missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Nitro-4-(trifluoromethyl)benzyl chloride, 98%, Thermo Scientific™

Product Code. 11330096
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11330096 1 g 1g
11320096 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11330096 Supplier Alfa Aesar™ Supplier No. H32191.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 225656-59-7
Molecular Formula C8H5ClF3NO2
Molecular Weight (g/mol) 239.578
MDL Number MFCD01631694
InChI Key JWFRBTUSUAFGOS-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl benzyl chloride, 1-chloromethyl-2-nitro-4-trifluoromethyl benzene, 4-chloromethyl-3-nitrobenzotrifluoride, 1-chloromethyl-2-nitro-4-trifluoromethyl-benzene, benzene,1-chloromethyl-2-nitro-4-trifluoromethyl
PubChem CID 2775786
IUPAC Name 1-(chloromethyl)-2-nitro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])CCl

Specifications

Melting Point 42°C to 45°C
Quantity 1 g
UN Number UN3261
Formula Weight 239.58
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)benzyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.