missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)phenol, 98%

Product Code. 11362886
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11362886 5 g 5g
11352886 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362886 Supplier Thermo Scientific Alfa Aesar Supplier No. L10604.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 400-99-7 | C7H4F3NO3 | 207.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 400-99-7
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00009791
InChI Key XZEDEVRSUANQEM-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl phenol, 4-hydroxy-3-nitrobenzotrifluoride, phenol, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethylphenol, 2-nitro-alpha,alpha,alpha-trifluoro-p-cresol, 4-hydroxy-3-nitrobenzotrifluoroide, p-cresol, 2-nitro-alpha,alpha,alpha-trifluoro, 2-nitro-4-trifluoromethyl-phenol, pubchem4150, 4-06-00-02150 beilstein handbook reference
PubChem CID 9816
IUPAC Name 2-nitro-4-(trifluoromethyl)phenol
SMILES OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F

Specifications

Density 1.473
Boiling Point 92°C to 94°C (12 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.504
Quantity 5 g
Beilstein 2215510
Formula Weight 207.11
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.