missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-5-(trifluoromethyl)aniline, TRC

Product Code. 30482047
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30482047 500 mg
30471172 1 g
30464759 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30482047 Supplier Toronto Research Chemicals Supplier No. TRCN587390500MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 402-14-2
Molecular Formula C7 H5 F3 N2 O2
Molecular Weight (g/mol) 206.122
Synonym alpha,alpha,alpha-Trifluoro-6-nitro-m-toluidine, 2-Nitro-5-(trifluoromethyl)aniline, 3-Amino-4-nitrobenzotrifluoride, 5-Trifluoromethyl-2-nitroaniline, NSC 10324, 2-Nitro-5-(trifluoromethyl)-benzenamine
IUPAC Name 2-nitro-5-(trifluoromethyl)aniline
SMILES Nc1cc(ccc1[N+](=O)[O-])C(F)(F)F

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H5F3N2O2/c8-7(9,10)4-1-2-6(12(13)14)5(11)3-4/h1-3H,11H2
Formula Weight 206.03
Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.