missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-m-xylene, 99%

Product Code. 11397485
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397485 100 g 100g
11307495 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397485 Supplier Thermo Scientific Alfa Aesar Supplier No. L05546.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 81-20-9 | C8H9NO2 | 151.165 g/mol

2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

Solubility
Miscible with ethanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-20-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007181
InChI Key HDFQKJQEWGVKCQ-UHFFFAOYSA-N
Synonym 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134
PubChem CID 6672
IUPAC Name 1,3-dimethyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)C)[N+](=O)[O-]

Specifications

Melting Point 14°C to 16°C
Density 1.11
Boiling Point 225°C to 227°C
Flash Point 87°C (188°F)
Refractive Index 1.522
Quantity 100 g
UN Number UN1665
Beilstein 2046066
Solubility Information Miscible with ethanol. Immiscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 2-Nitro-m-xylene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.