missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetanilide, 98+%

Product Code. 11432668
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432668 25 g 25g
11422668 5 g 5g
11442668 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11432668 Supplier Thermo Scientific Alfa Aesar Supplier No. A16677.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 552-32-9 | C8H8N2O3 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-32-9
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00016991
InChI Key BUNFNRVLMKHKIT-UHFFFAOYSA-N
Synonym n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
PubChem CID 11090
IUPAC Name N-(2-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 90°C to 93°C
Density 1.42
Quantity 25 g
Beilstein 1959178
Merck Index 14,6581
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material 2'-Nitroacetanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.