missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonamide, 96%, ACROS Organics™

Product Code. 10656174 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10656174 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10656174 Supplier Thermo Scientific Acros Supplier No. 211790250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 5455-59-4
CAS Min % 95.0
CAS Max % 100.0
Melting Point 189°C to 194°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C6H6N2O4S
Linear Formula O2NC6H4SO2NH2
MDL Number MFCD00009807
Quantity 25 g
Beilstein 11, 68
Synonym 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n
InChI Key GNDKYAWHEKZHPJ-UHFFFAOYSA-N
SMILES NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O
IUPAC Name 2-nitrobenzene-1-sulfonamide
Molecular Weight (g/mol) 202.18
PubChem CID 138510
Formula Weight 202.18
Percent Purity ≥95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzenesulfonamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.