missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonamide, 97+%

Product Code. 11487637
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11487637 5 g 5g
11369569 10 g 10g
11497637 25 g 25g
11407647 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11487637 Supplier Thermo Scientific Alfa Aesar Supplier No. A15689.06
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 5455-59-4 | C6H6N2O4S | 202.18 g/mol

It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5455-59-4
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00009807
InChI Key GNDKYAWHEKZHPJ-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n
PubChem CID 138510
IUPAC Name 2-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 188°C to 194°C
Quantity 5 g
Beilstein 2214215
Solubility Information Slightly soluble in water.
Formula Weight 202.19
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzenesulfonamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.