missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic acid, 95%, Thermo Scientific Chemicals

Product Code. 10617631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10617631 250 g Glass bottle 250g
10607631 5 g Glass bottle 5g
10453215 25 g Glass bottle 25g
10006003 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10617631 Supplier Thermo Scientific Acros Supplier No. 278322500
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 552-16-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-16-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007137
InChI Key SLAMLWHELXOEJZ-UHFFFAOYSA-N
Synonym 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
PubChem CID 11087
ChEBI CHEBI:25620
SMILES C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 85.0
CAS Max % 100.0
Melting Point 141.0°C to 148.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COOH
Quantity 250 g
Beilstein 09, 370
Fieser 13,231
Merck Index 14, 6589
Solubility Information Solubility in water: 6.8g/l. Other solubilities: 1g/3mL alcohol- 1g/2.5mL acetone,1g/220mL chloroform- 1g/2.5mL methanol,1g/4.5mL ether,slightly soluble in benzene,cs2,petroleum ether
Formula Weight 167.12
Percent Purity 95%
Physical Form Fine Powder
Chemical Name or Material 2-Nitrobenzoic acid, 85-90%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.