missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic acid, 95%

Product Code. p-172802 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
5g
This item is not returnable. View return policy

Product Code. 10607631

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 552-16-9 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-16-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007137
InChI Key SLAMLWHELXOEJZ-UHFFFAOYSA-N
Synonym 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
PubChem CID 11087
ChEBI CHEBI:25620
SMILES C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 85.0
CAS Max % 100.0
Melting Point 141.0°C to 148.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4COOH
Quantity 5 g
Beilstein 09, 370
Fieser 13,231
Merck Index 14, 6589
Solubility Information Solubility in water: 6.8g/l. Other solubilities: 1g/3mL alcohol- 1g/2.5mL acetone,1g/220mL chloroform- 1g/2.5mL methanol,1g/4.5mL ether,slightly soluble in benzene,cs2,petroleum ether
Formula Weight 167.12
Percent Purity 95%
Physical Form Fine Powder
Chemical Name or Material 2-Nitrobenzoic acid, 85-90%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2-Nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.