missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl bromide, 97%

Product Code. 10153480 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10153480 5 g Glass bottle 5g
10408110 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10153480

Brand: Thermo Scientific Acros 148310050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3958-60-9 | C7H6BrNO2 | 216.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3958-60-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00007184
InChI Key HXBMIQJOSHZCFX-UHFFFAOYSA-N
Synonym 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
PubChem CID 77569
IUPAC Name 1-(bromomethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CBr)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 44°C to 48°C
Color Yellow to Brown
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula O2NC6H4CH2Br
Quantity 5 g
Beilstein 05, I, 164
Solubility Information Solubility in water: insoluble
Formula Weight 216.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Nitrobenzyl bromide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Nitrobenzyl bromide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.