missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl bromide, 98+%

Product Code. 11444484
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444484 5 g 5g
11454484 25 g 25g
11464484 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444484 Supplier Thermo Scientific Alfa Aesar Supplier No. A13127.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3958-60-9 | C7H6BrNO2 | 216.034 g/mol

2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzyl bromide is used in the preparation of S-2-nitrobenzyl-cysteine by reaction with L-cysteine. It is also used for the introduction of the 2-nitrobenzyl protecting group in organic synthesis. It plays an important role in the production of expectorant agent. Further, it is employed for caging unprotected cysteine-containing or thiophosphorylated peptides in aqueous solution. In addition to this, it is used in the preparation of (R)- and (S)-3-amino-3,4-dihydro-1H-quinolin-2-one.

Solubility
Insoluble in water.

Notes
Moisture sensitive. Incompatible with alcohols, bases, amines and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3958-60-9
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00007184
InChI Key HXBMIQJOSHZCFX-UHFFFAOYSA-N
Synonym 2-nitrobenzyl bromide, 1-bromomethyl-2-nitrobenzene, o-nitrobenzyl bromide, alpha-bromo-2-nitrotoluene, 2-bromomethyl nitrobenzene, 1-bromomethyl-2-nitro-benzene, benzene, 1-bromomethyl-2-nitro, ccris 7966, 2-nitro benzyl bromide, 2-nitro-benzyl bromide
PubChem CID 77569
IUPAC Name 1-(bromomethyl)-2-nitrobenzene
SMILES C1=CC=C(C(=C1)CBr)[N+](=O)[O-]

Specifications

Melting Point 45°C to 48°C
Flash Point >110°C (230°F)
Quantity 5 g
UN Number UN3261
Beilstein 638991
Solubility Information Insoluble in water.
Formula Weight 216.04
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.