missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrofluorene, 98%

Product Code. 15462078
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462078 5 g 5g
15472078 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462078 Supplier Thermo Scientific Alfa Aesar Supplier No. H55744.06
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 607-57-8 | C13H9NO2 | 211.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 607-57-8
Molecular Formula C13H9NO2
Molecular Weight (g/mol) 211.22
MDL Number MFCD00001117
InChI Key XFOHWECQTFIEIX-UHFFFAOYSA-N
Synonym 2-nitrofluorene, nitrofluorene, 9h-fluorene, 2-nitro, fluorene, 2-nitro, unii-191ll4u4gz, ccris 1189, dsstox_cid_971, dsstox_rid_75898, dsstox_gsid_20971
PubChem CID 11831
ChEBI CHEBI:1224
IUPAC Name 2-nitro-9H-fluorene
SMILES [O-][N+](=O)C1=CC=C2C(CC3=CC=CC=C23)=C1

Specifications

Melting Point 156°C to 158°C
Color White to Brown
Quantity 5 g
UN Number UN3077
Formula Weight 211.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Nitrofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.