missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrophenol, ≥99% (HPLC), For Environmental Analysis, Honeywell™ Fluka™

Product Code. 15663970 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
Unit Size:
25g
This item is not returnable. View return policy

Product Code. 15663970

Brand: Honeywell-Fluka 3344425G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

for environmental analysis,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 88-75-5
Melting Point 45°C
Color Yellow
Boiling Point 214°C
Flash Point 102°C (215.6°F)
Infrared Spectrum Conforms to structure
Packaging Glass Bottle
Molecular Formula C6H5NO3
Linear Formula O2NC6H4OH
MDL Number MFCD00011688
Quantity 25 g
UN Number UN1663
Beilstein 775403
Synonym o-nitrophenol, 2-hydroxynitrobenzene, phenol, 2-nitro, o-hydroxynitrobenzene, phenol, o-nitro, nitrophenol, o-nitrofenol, ortho-nitrophenol, phenol, nitro, o-nitrofenol czech
InChI Key IQUPABOKLQSFBK-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1[N+]([O-])=O
IUPAC Name 2-nitrophenol
Molecular Weight (g/mol) 139.11
PubChem CID 6947
ChEBI CHEBI:16260
Formula Weight 139.11g/mol
Percent Purity ≥99% (HPLC)
Chemical Name or Material 2-Nitrophenol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.