missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroresorcinol, 98%

Product Code. p-7020869
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11459833

Brand: Thermo Scientific Alfa Aesar A12970.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 601-89-8 | C6H5NO4 | 155.11 g/mol

It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of porous materials by 2-nitroresorcinol/cyanuric chloride thermal polycondensation in emulsions. It is used as an active pharmaceutical intermediate and an intermediate for organic synthesis.

Solubility
Slightly soluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 601-89-8
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00007124
InChI Key ZLCPKMIJYMHZMJ-UHFFFAOYSA-N
Synonym 2-nitroresorcinol, 2-nitrobenzene-1,3-diol, 1,3-benzenediol, 2-nitro, resorcinol, 2-nitro, 2-nitro-1,3-benzenediol, 1,3-dihydroxy-2-nitrobenzene, 1,3-benzenediol, nitro, 2,6-dihydroxynitrobenzene, nitroresorcinol, acmc-209mid
PubChem CID 11760
IUPAC Name 2-nitrobenzene-1,3-diol
SMILES OC1=CC=CC(O)=C1[N+]([O-])=O

Specifications

Melting Point 81°C to 85°C
Quantity 5 g
UN Number UN2811
Beilstein 2048819
Solubility Information Slightly soluble in water. Solubility in methanol is almost transparent.
Formula Weight 155.11
Percent Purity 98%
Chemical Name or Material 2-Nitroresorcinol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.