missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroterephthalic acid 1-methyl ester, 97%

Product Code. 11436220
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11436220 5 g 5g
11446220 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11436220 Supplier Thermo Scientific Alfa Aesar Supplier No. H27121.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 35092-89-8 | C9H7NO6 | 225.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35092-89-8
Molecular Formula C9H7NO6
Molecular Weight (g/mol) 225.16
MDL Number MFCD00024510
InChI Key MIIADZYPHVTLPR-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl-3-nitrobenzoic acid, 1-methyl 2-nitroterephthalate, methyl hydrogen 2-nitroterephthalate, 3-nitro-4-carbomethoxybenzoic acid, 2-nitroterephthalic acid 1-methyl ester, methyl 2-nitro-4-carboxybenzoate, methyl-2-nitro-4-carboxy-benzoate, 4-carbomethoxy-3-nitro-benzoic acid
PubChem CID 98592
IUPAC Name 4-(methoxycarbonyl)-3-nitrobenzoic acid
SMILES COC(=O)C1=CC=C(C=C1[N+]([O-])=O)C(O)=O

Specifications

Melting Point 173°C to 178°C
Quantity 5 g
Formula Weight 225.17
Percent Purity 97%
Chemical Name or Material 2-Nitroterephthalic acid 1-methyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.