missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrotoluene, 99+%

Product Code. 11465503
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11465503 100 g 100g
11475503 500 g 500g
11485503 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11465503 Supplier Thermo Scientific Alfa Aesar Supplier No. A13446.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Used as an intermediate, reagent, as a precursor and as a nitrogen, carbon, energy supplement | CAS: 88-72-2 | C7H7NO2 | 137.138 g/mol

2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.

Solubility
Miscible with acetone, benzene, chloroform, diethyl ether, ethanol and petroleum ether. Immiscible with water.

Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, sulfuric acid, reducing agents, hydrogen and sodium.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007157
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzene
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]

Specifications

Melting Point -4°C
Density 1.166
Boiling Point 224°C to 226°C
Flash Point 106°C (222°F)
Refractive Index 1.545
Quantity 100 g
UN Number UN1664
Beilstein 1907580
Merck Index 14,6650
Solubility Information Miscible with acetone,benzene,chloroform,diethyl ether,ethanol and petroleum ether; Immiscible with water.
Formula Weight 137.14
Percent Purity ≥99%
Chemical Name or Material 2-Nitrotoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Nitrotoluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: 2-Nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.