missing translation for 'onlineSavingsMsg'
Learn More

2-(p-Tolyl)propionic Acid, TRC

Product Code. 30136841
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136841 5 g 5g
30008197 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136841 Supplier Toronto Research Chemicals Supplier No. TRCT5367805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 938-94-3
Molecular Formula C10 H12 O2
Molecular Weight (g/mol) 164.2
Synonym (2RS)-2-(4-Methylphenyl)propanoic acid, Ibuprofen Imp. D (EP), Benzeneacetic acid, α,4-dimethyl-, Hydratropic acid, p-methyl- (6CI,7CI,8CI), α,4-Dimethylbenzeneacetic acid, (±)-2-(p-Methylphenyl)propionic acid, 2-(4-Methylphenyl)propanoic acid, 2-(4-Methylphenyl)propionic acid, 2-(p-Methylphenyl)propionic acid, 2-p-Tolylpropionic acid, p-Methylhydratropic acid, α-(p-Tolyl)propionic acid
IUPAC Name 2-(4-methylphenyl)propanoic acid
SMILES CC(C(=O)O)c1ccc(C)cc1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C10H12O2/c1-7-3-5-9(6-4-7)8(2)10(11)12/h3-6,8H,1-2H3,(H,11,12)
Formula Weight 164.0837
Percent Purity >95%
Chemical Name or Material (2RS)-2-(4-Methylphenyl)propanoic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.