missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyaniline, 98%

Product Code. 11337495
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337495 25 g 25g
11347495 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337495 Supplier Thermo Scientific Alfa Aesar Supplier No. L05558.14
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2688-84-8 | C12H11NO | 185.226 g/mol

2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenoxyaniline serves as an antiinflammatory agent that inhibits preferentially COX-2 over COX-1. It is used in the preparation of sodium primary amide complex, 4-methyl-2-(2-phenoxyphenyl)azo-phenol and 2-acetoaminodiphenyl ether. Further, it is used to form complexes with beta-cyclodextrin.

Solubility
Slightly soluble in chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00035765
InChI Key NMFFUUFPJJOWHK-UHFFFAOYSA-N
Synonym 2-aminodiphenyl ether, o-phenoxyaniline, 2-aminophenyl phenyl ether, benzenamine, 2-phenoxy, 2-ade, 2-phenoxybenzenamine, o-aminophenyl phenyl ether, aniline, 2-phenoxy, aniline, o-phenoxy, 2-phonoxyaniline
PubChem CID 75899
IUPAC Name 2-phenoxyaniline
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2N

Specifications

Melting Point 46°C to 48°C
Boiling Point 170°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 777766
Solubility Information Slightly soluble in chloroform and methanol.
Formula Weight 185.23
Percent Purity 99%
Chemical Name or Material 2-Phenoxyaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.