missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzaldehyde, 98%, Thermo Scientific™

Product Code. 11325489
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325489 1 g 1g
11315489 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11325489 Supplier Thermo Scientific Alfa Aesar Supplier No. H50448.03
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Phenoxybenzaldehyde is used as a reagent in the synthesis of chiral 1,3,4-oxadiazol-2-ones as potent and highly selective FAAH inhibitors. Also used as a reagent in the synthesis of [4-(phenoxy)pyridin-3-yl]methylamines; a new class of selective noradrenaline reuptake inhibitors (NRI).

Solubility
Very slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19434-34-5
Molecular Formula C13H10O2
Molecular Weight (g/mol) 198.22
MDL Number MFCD00800666
InChI Key IMPIIVKYTNMBCD-UHFFFAOYSA-N
Synonym o-phenoxybenzaldehyde, 2-phenoxybenzenecarbaldehyde, 2-formyldiphenyl ether, phenoxybenzaldehyde, 2-phenoxy-benzaldehyde, pubchem16236, phenoxybenzenecarbaldehyde, opera_id_1384
PubChem CID 88060
IUPAC Name 2-phenoxybenzaldehyde
SMILES O=CC1=CC=CC=C1OC1=CC=CC=C1

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Very slightly soluble in water.
Formula Weight 198.22
Percent Purity 98%
Chemical Name or Material 2-Phenoxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.