missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxybenzoyl chloride, TECH, Thermo Scientific™

Product Code. 10121773
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10121773 1 g 1g
10758455 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10121773 Supplier Thermo Scientific Maybridge Supplier No. CC49902DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 40501-36-8
Molecular Formula C13H9ClO2
Molecular Weight (g/mol) 232.663
MDL Number MFCD03424711
InChI Key BMGKQFRMINVVPP-UHFFFAOYSA-N
Synonym benzoylchloride, 2-phenoxy, phenoxybenzoyl chloride, 2-phenoxy benzoyl chloride, 2-phenoxy-benzoyl chloride, benzoyl chloride, 2-phenoxy, 2-phenoxybenzoic acid chloride, 2-chlorocarbonyl diphenyl ether
PubChem CID 11075303
IUPAC Name 2-phenoxybenzoyl chloride
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2C(=O)Cl

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Quantity 1 g
Formula Weight 232.67
Percent Purity 90%
Chemical Name or Material 2-Phenoxybenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.