missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, 99%

Product Code. 10511571 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass bottle
Metal drum
Plastic drum
Unit Size:
1 L
2.5 L
25 L
250mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10511571 1 L Glass bottle 1 L
10367010 250 mL Glass bottle 250mL
10043763 2.5 L Glass bottle 2.5 L
10176223 5 L Plastic drum 5 L
11944731 25 L Metal drum 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10511571 Supplier Thermo Scientific Acros Supplier No. 130240010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C | CAS: 122-99-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002857
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Colorless
Density 1.1000g/mL
Boiling Point 237.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5OCH2CH2OH
Refractive Index 1.5360 to 1.5400
Quantity 1 L
Beilstein 06,146
Merck Index 15,7369
Specific Gravity 1.1
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: freely soluble in alcohol and ether,miscible with acetone and ethyl acetate
Viscosity 29 mPa.s (20°C)
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Phenoxyethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Ethylene glycol monophenyl ether

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.