missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxyethanol, 99%

Product Code. 11944731 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
5 L
25 L
Packaging:
Glass bottle
Metal drum
Plastic drum
Unit Size:
1 L
2.5 L
25 L
250mL
5 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11944731 25 L Metal drum 25 L
10367010 250 mL Glass bottle 250mL
10511571 1 L Glass bottle 1 L
10043763 2.5 L Glass bottle 2.5 L
10176223 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11944731 Supplier Thermo Scientific Acros Supplier No. 130240250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Phenoxyethanol, 99%, C8H10O2, CAS Number-122-99-6, ethylene glycol monophenyl ether, ethanol, 2-phenoxy, 1-hydroxy-2-phenoxyethane, phenoxyethanol, phenoxyethyl alcohol, phenoxethol, phenoxytol, phenoxetol, phenyl cellosolve, ethylene glycol phenyl ether, 25L, 98.5% min. (GC), 06,146, 237.0 deg.C | CAS: 122-99-6 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122-99-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002857
InChI Key QCDWFXQBSFUVSP-UHFFFAOYSA-N
Synonym phenoxyethanol, ethylene glycol monophenyl ether, phenyl cellosolve, ethanol, 2-phenoxy, phenoxytol, phenoxethol, phenoxetol, ethylene glycol phenyl ether, phenoxyethyl alcohol, 1-hydroxy-2-phenoxyethane
PubChem CID 31236
ChEBI CHEBI:64275
IUPAC Name 2-phenoxyethanol
SMILES C1=CC=C(C=C1)OCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 11.0°C to 13.0°C
Color Colorless
Density 1.1000g/mL
Boiling Point 237.0°C
Flash Point 130°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Linear Formula C6H5OCH2CH2OH
Refractive Index 1.5360 to 1.5400
Quantity 25 L
Beilstein 06,146
Merck Index 15,7369
Specific Gravity 1.1
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: freely soluble in alcohol and ether,miscible with acetone and ethyl acetate
Viscosity 29 mPa.s (20°C)
Formula Weight 138.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Phenoxyethanol
Show More Show Less
Hazard Category
  • Specific target organ toxicity Category 3
Hazard Statement
  • H335-May cause respiratory irritation.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.