missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxylaniline, TRC

Product Code. 30134574
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134574 25 g 25g
30133047 100 g 100g
17962285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134574 Supplier Toronto Research Chemicals Supplier No. TRCP29820025G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2688-84-8
Molecular Formula C12 H11 N O
Molecular Weight (g/mol) 185.22
Synonym 2-Phenoxyaniline, Nimesulide Imp. C (EP), Benzenamine, 2-phenoxy-, Aniline, o-phenoxy- (6CI,7CI,8CI), 2-Phenoxybenzenamine, 2-Aminodiphenyl ether, 2-Aminophenyl phenyl ether, 2-Phenoxaniline, 2-Phenoxylaniline, 2-Phenoxyphenylamine, NSC 39655, o-Aminodiphenyl ether, o-Aminophenyl phenyl ether, o-Phenoxyaniline
IUPAC Name 2-phenoxyaniline
SMILES Nc1ccccc1Oc2ccccc2

Specifications

Quantity 25 g
InChI Formula InChI=1S/C12H11NO/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9H,13H2
Formula Weight 185.0841
Percent Purity >95%
Chemical Name or Material 2-Phenoxyaniline
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.