missing translation for 'onlineSavingsMsg'
Learn More

2-Phenoxylaniline, TRC

Product Code. 17962285
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17962285 5 g 5g
30134574 25 g 25g
30133047 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17962285 Supplier Toronto Research Chemicals Supplier No. TRCP2982005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2688-84-8
Formula Weight 185.0841
InChI Formula InChI=1S/C12H11NO/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9H,13H2
IUPAC Name 2-phenoxyaniline
Molecular Formula C12 H11 N O
Molecular Weight (g/mol) 185.22
Percent Purity >95%
SMILES Nc1ccccc1Oc2ccccc2
Synonym 2-Phenoxyaniline, Nimesulide Imp. C (EP), Benzenamine, 2-phenoxy-, Aniline, o-phenoxy- (6CI,7CI,8CI), 2-Phenoxybenzenamine, 2-Aminodiphenyl ether, 2-Aminophenyl phenyl ether, 2-Phenoxaniline, 2-Phenoxylaniline, 2-Phenoxyphenylamine, NSC 39655, o-Aminodiphenyl ether, o-Aminophenyl phenyl ether, o-Phenoxyaniline
Chemical Name or Material 2-Phenoxyaniline
Grade HPLC
Show More Show Less

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.