missing translation for 'onlineSavingsMsg'
Learn More

[2-(Phenoxymethyl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10682695
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10682695 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10682695 Supplier Thermo Scientific Maybridge Supplier No. CC58509DA
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 34904-98-8
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C14H14O2
MDL Number MFCD08690312
Quantity 1 g
Synonym 2-phenoxymethyl phenyl methanol, benzenemethanol,2-phenoxymethyl, 2-phenoxymethyl phenyl methan-1-ol
InChI Key RMRTWBZKENSMCI-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)OCC2=CC=CC=C2CO
IUPAC Name [2-(phenoxymethyl)phenyl]methanol
Molecular Weight (g/mol) 214.264
PubChem CID 22932170
Formula Weight 214.26
Percent Purity 97%
Chemical Name or Material [2-(Phenoxymethyl)phenyl]methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.