missing translation for 'onlineSavingsMsg'
Learn More

(2-Phenoxyphenyl)methanol, ≥97%, Thermo Scientific™

Product Code. 10317454
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10317454 250 mg 250mg
10082264 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317454 Supplier Thermo Scientific Maybridge Supplier No. CC49909CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 13807-84-6
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00017297
InChI Key VMZBMTWFHYYOIN-UHFFFAOYSA-N
Synonym 2-phenoxyphenyl methanol, 2-phenoxybenzyl alcohol, o-phenoxybenzyl alcohol, phenoxyphenylmethanol, 2-phenoxybenzylalcohol, 2-phenoxybenzenemethanol, o-phenoxy benzyl alcohol, 2-phenoxy-benzenemethanol, benzenemethanol, phenoxy, 2-phenoxy-phenyl-methanol
PubChem CID 3660111
IUPAC Name (2-phenoxyphenyl)methanol
SMILES C1=CC=C(C=C1)OC2=CC=CC=C2CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250 mg
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material (2-Phenoxyphenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.