missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 108238-09-1 |
| Melting Point | 110°C to 112°C |
| Color | White |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94% min. (HPLC) |
| Molecular Formula | C12H11BO3 |
| MDL Number | MFCD01001592 |
| Quantity | 10 g |
| Synonym | 2-phenoxybenzeneboronic acid, 2-phenoxyphenyl boronic acid, 2-phenoxyphenyl boranediol, 2-phenoxy phenylboronic acid, boronic acid, 2-phenoxyphenyl, pubchem9521, acmc-1bnrk, ksc174e3r, 2-phenoxy-phenyl boronic acid, 2-phenoxyphenylboronicacid |
| InChI Key | AVOWPOFIQZSVGV-UHFFFAOYSA-N |
| Vedi altri risultati |
Sicurezza e movimentazione
Identificativo prodotto
- 2-Phenoxyphenylboronic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?