missing translation for 'onlineSavingsMsg'
Learn More

2-Phenyl-2-butenal, (E)+(Z), 97%

Product Code. 11412257
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412257 5 g 5g
11422257 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412257 Supplier Thermo Scientific Alfa Aesar Supplier No. A14185.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4411-89-6 | C10H10O | 146.19 g/mol

2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Phenyl-2-butenal is used as a food additive. It is also used as intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in oil and ethanol. Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4411-89-6
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00053158
InChI Key DYAOGZLLMZQVHY-MBXJOHMKSA-N
Synonym 2-phenyl-2-butenal, 2-phenylcrotonaldehyde, 2-phenylbutenal, alpha-phenylcrotonaldehyde, unii-n8emo6yrv9, e-2-phenylbut-2-enal, 2-butenal, 2-phenyl, alpha-ethylidenebenzeneacetaldehyde, n8emo6yrv9, e-2-phenyl-2-butenal
PubChem CID 6429333
IUPAC Name (E)-2-phenylbut-2-enal
SMILES C\C=C(\C=O)C1=CC=CC=C1

Specifications

Boiling Point 115°C (15 mmHg)
Refractive Index 1.561
Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Soluble in oil and ethanol. Insoluble in water.
Formula Weight 146.19
Percent Purity 97%
Chemical Name or Material 2-Phenyl-2-butenal, (E)+(Z)
compliance-icons
Product Identifier
  • 2-Phenyl-2-butenal,(E)+(Z)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Phenyl-2-butenal,(E)+(Z)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.