missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-nitrobenzotrifluoride, 98+%

Product Code. 11493058
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11493058

Brand: Thermo Scientific Alfa Aesar A18780.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 393-09-9 | C7H3F4NO2 | 209.1 g/mol

5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoro-2-nitrobenzotrifluoride may be used as monomer in the synthesis of hyperbranched poly(aryl ether). Also used in the preparation of androgen receptor modulators and other pharmaceutical compounds.

Solubility
Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-09-9
Molecular Formula C7H3F4NO2
Molecular Weight (g/mol) 209.1
MDL Number MFCD00039741
InChI Key WMQOSURXFLBTPC-UHFFFAOYSA-N
Synonym 5-fluoro-2-nitrobenzotrifluoride, 4-fluoro-1-nitro-2-trifluoromethyl benzene, 4-fluoro-2-trifluoromethyl nitrobenzene, 2-nitro-alpha,alpha,alpha,5-tetrafluorotoluene, 2-nitro-5-fluorobenzotrifluoride, 1-fluoro-3-trifluoromethyl-4-nitrobenzene, 2-trifluoromethyl-4-fluoronitrobenzene, benzene, 4-fluoro-1-nitro-2-trifluoromethyl, 4-fluoro-1-nitro-2-trifluoromethyl-benzene, 2-nitro-5-fluorotrifluorotoluol
PubChem CID 223094
IUPAC Name 4-fluoro-1-nitro-2-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1F)C(F)(F)F)[N+](=O)[O-]

Specifications

Melting Point 22°C to 24°C
Density 1.497
Boiling Point 87°C to 88°C (2 mmHg)
Flash Point 88°C (190°F)
Refractive Index 1.46
Quantity 5 g
UN Number UN2811
Beilstein 3304470
Solubility Information Soluble in chloroform and methanol.
Formula Weight 209.1
Percent Purity ≥98%
Chemical Name or Material 5-Fluoro-2-nitrobenzotrifluoride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.